Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Multiparameter PH/EC/DO Meter | Avantor

Multiparameter PH/EC/DO Meter

Multiparameter PH/EC/DO meter
Catalog # CA470349-854
Supplier:   Hanna
Fournisseur #:  HI98194
Multiparameter PH/EC/DO Meter
Catalog # CA470349-854
Supplier:  Hanna
Multiparameter PH/EC/DO Meter
Multiparameter PH/EC/DO Meter

Multiparameter PH/EC/DO meter

Price
$2,193.39
Log in pour la disponibilité
Each
Product Details & Documents

This multiparameter meter is a rugged, waterproof multiparameter meter. This meter's multi-sensor probe allows for the measurement of key parameters including pH, ORP, conductivity, dissolved oxygen, and temperature. The multiparameter probe transmits readings digitally to the meter, where data points can be displayed and logged. The complete water quality meter is simple to setup.

  • Comes as a complete system, allowing you to start taking measurements as soon as you receive it
  • Results are trustworthy every time
  • Great for environmental and industrial professionals
  • Color coded, field replaceable sensors
  • Quick connect
  • IP 68 rating
  • Rugged carrying case

Backlit Graphic LCD Display - This meter features a backlit graphic LCD with on-screen help and the capability to display up to twelve parameters simultaneously. The graphic display allows for the use of virtual keys to provide for an intuitive user interface.

Waterproof Protection - The multiparameter meter is enclosed in an IP 67 rated waterproof casing and can withstand immersion in water at a depth of 1m for up to 30 minutes. The probe features an IP 68 rating for continuous immersion in water. Quick Connect Digital Probe - The probe features a Quick Connect DIN connector that makes a waterproof connection with the meter.

Color Coded, Field Replaceable Sensors - Sensor replacement is quick and easy with field replaceable, screw type connectors that are color-coded for easy sensor port identification. Auto-sensor Recognition - The probe and meter automatically recognize the sensors that are connected. Any ports not used on the probe will not have the parameter displayed or be configurable. Automatic Temperature Compensation - Integrated temperature sensor allows for automatic temperature compensation of pH, conductivity, and dissolved oxygen measurements. Automatic Barometric Pressure Compensation - The meter features a built-in barometer with user-selectable units for dissolved oxygen pressure compensation.

Standard or Quick Calibration - Quick Calibration provides a speedy, single point calibration for pH, conductivity, and dissolved oxygen. Standard pH calibration options are available for calibrating up to three points from a selection of five standard buffers and one custom buffer. Conductivity calibration is a single point from six standard selections or one custom standard. Dissolved oxygen calibration is up to two standard points or a single custom point. GLP Data - This meter includes a GLP feature that allows users to view calibration data and expiration information at the touch of a key. Calibration data includes date, time, buffers/standards used for calibration, and slope characteristics. Data Logging - This meter allows users to store up to 45,000 continuous or log-on-demand samples with logging intervals from one second to three hours.

Intuitive Keypad - The fitted rubber keypad has dedicated keys for power, backlight, up/down arrows, help and alphanumeric characters. The meter also features two virtual soft keys that navigate the user through the configuration of each parameter, meter setup, and logging of data. The interface is intuitive for any user’s level of experience. Dedicated Help Key - Contextual help is always available through a dedicated ‘HELP’ key. Clear tutorial messages and directions are available on-screen to quickly guide users through setup and calibration. The help information displayed is relative to the setting/option being viewed.

PC Connectivity - Logged data can be transferred to a Window’s compatible PC with the included micro-USB cable and software. Long Battery Life - The display of the water quality meter has a battery icon indicator to show the remaining power. The meter uses four 1.5 V AA batteries that provide up to 360 hours of battery life. Rugged Thermoformed Carrying Case - The meter, probe, and all accessories are supplied in a rugged carrying case designed to provide years of use. The inside compartment of the carrying case is thermoformed to securely hold and protect all the components.

Ordering information: Replaceable sensors are purchased separately. Calibration solutions, pH storage solutions, and pH cleaning solutions are purchased seperately. Software can also be requested. Micro USB cable can be purchased separately as well. Contact your sales representative for more information.

Spécifications
  • Désignation:
    Multiparameter PH/EC/DO meter
  • Operating temperature range:
    0...50 °C (32...122 °F) [temperature of sample]
  • mV range:
    ±2000.0 mV (ORP)
  • DO range:
    0.0 - 500.0%, 0.00 - 50.00 ppm (mg/L)
  • pH range:
    0.00 - 14.00 pH
  • Conductivity resolution:
    Manual: 1 µS/cm, 0.001 mS/cm, 0.01 mS/cm, 0.1 mS/cm, 1 mS/cm
    Automatic: 1 µS/cm from 0 - 9999 µS/cm, 0.01 mS/cm from 10.00 - 99.99 mS/cm, 0.1 mS/cm from 100.0 - 400.0 mS/cm
  • DO accuracy:
    0.0 - 300.0%: ±1.5% of reading or ±1.0% whichever is greater
    300.0 - 500.0%: ±3% of reading 0.00 - 30.00 ppm (mg/L): ±1.5% of reading or ±0.10 ppm (mg/L) whichever is greater
    30.00 ppm (mg/L) - 50.00 ppm (mg/L): ±3% of reading
  • Calibration:
    pH: automatic one, two, or three points with automatic recognition of five standard buffers (pH 4.01, 6.86, 7.01, 9.18, 10.01) or one custom buffer
    ORP: automatic at one custom point (relative mV)
    Conductivity: automatic single point, with six standard solutions (84 µS/cm, 1413 µS/cm, 5.00 mS/cm, 12.88 mS/cm, 80.0 mS/cm, 111.8 mS/cm) or custom point
    TDS: based on conductivity calibration
    Dissolved oxygen: automatic one or two points at 0 and 100% or one custom point
  • mV resolution:
    0.1 mV (ORP)
  • TDS resolution:
    Manual: 1 ppm (mg/L), 0.001 ppt (g/L), 0.01 ppt (g/L), 0.1 ppt (g/L), 1 ppt (g/L)
    Automatic: 1 ppm (mg/L) from 0 - 9999 ppm (mg/L), 0.01 ppt (g/L) from 10.00 - 99.99 ppt (g/L), 0.1 ppt (g/L) from 100.0 - 400.0 ppt (g/L)
    Automatic ppt (g/L): 0.001 ppt (g/L) from 0.000 - 9.999 ppt (g/L), 0.01 ppt (g/L) from 10.00 - 99.99 ppt (g/L), 0.1 ppt (g/L) from 100.0 - 400.0 ppt (g/L)
  • TDS accuracy:
    ±1% of reading or ±1 µS/cm, whichever is greater
  • Cat. No.:
    CA470349-854
  • Conductivity accuracy:
    ±1% of reading or ±1 µS/cm whichever is greater
  • pH accuracy:
    ±0.02 pH
  • pH resolution:
    0.01 pH
  • DO resolution:
    0.0 - 300.0%: ±1.5% of reading or ±1.0% whichever is greater
    300.0 - 500.0%: ±3% of reading 0.00 - 30.00 ppm (mg/L): ±1.5% of reading or ±0.10 ppm (mg/L) whichever is greater
    30.00 ppm (mg/L) - 50.00 ppm (mg/L): ±3% of reading
  • Conductivity range:
    0 - 200 mS/cm (absolute EC up to 400 mS/cm)
  • TDS range:
    0 - 400000 ppm (mg/L) (the maximum value depends on the TDS factor)
  • mV accuracy:
    ±1.0 mV (ORP)
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.