Transfer Caps for Storage Bottles, Square, Closed System Solution
Bi-directional transfer cap, 250 ml Square storage bottle closed system solution
Catalog # 77509-130
Supplier: NEST SCIENTIFIC
Fournisseur #: C50921-BHB060B
Transfer Caps for Storage Bottles, Square, Closed System Solution
Catalog # 77509-130
Supplier: NEST SCIENTIFIC
Jump to:
Product Details & Documents
These injection-molded transfer caps are highly compatible with other major brands.
Spécifications
- Désignation:Bi-directional transfer cap
- For Use With:250 ml Square storage bottle closed system solution