PinPoint™ Pipette Controller | Avantor
PinPoint™ Pipette Controller
PinPoint™ pipette controller
Catalog # 470356-092
Fournisseur #:  UNPINPNT01
PinPoint™ Pipette Controller
Catalog # 470356-092
PinPoint™ Pipette Controller
PinPoint™ Pipette Controller
PinPoint™ pipette controller
Price
$317.55
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Digital display makes transferring liquids easier.
  • Compatible with glass or plastic serological pipettes (0.1 to 100 ml)
  • LCD indicates battery charge and 6 speed levels
  • Fills a 25 ml pipette in less than 5 seconds
  • Lithium-ion battery charges fully within 3 hours and can provide up to 8 hours of use
Ultra-light and powerful pipette controller is designed for comfort as well as accuracy. Intuitive 6 level speed setting and sensitive triggers provide a wide range of aspirating/dispensing speeds. Pipette controller is ergonomically designed for extended usage.
Certifications: Meets the requirements of the applicable EU directives, UK regulations, UL, and CAN/CSA.
Ordering information: The PinPoint™ Pipette Controller comes with an AC adapter with USB connection, a 0.45 μm hydrophobic filter, a user manual, wall mount, and two-position desk stand.
Spécifications
  • Désignation:
    PinPoint™ pipette controller