Transfer Caps for High Efficient Erlenmeyer Flasks, Multifunction Liquid System
Sterile PETG square storage bottle bi-directional transfer cap with ¹/₄" TPE tube, female Luer connector with male sealing plug vent filter, 2 L Flasks, —, —
77509-122
: C501AA-BHC060B
Transfer Caps for High Efficient Erlenmeyer Flasks, Multifunction Liquid System
77509-122
Transfer caps for high efficient erlenmeyer flask.
- Compatible with 2/3/5 L high efficient Erlenmeyer flasks
- The liquid inlet tubing is provided with a PTFE needle filter, which solves the issue of liquid remaining in the tubing during the
- feeding process
Different from bi-directional liquid transfer cap, the multifunctional transfer cap can be directly placed in an incubator for culture after the liquid transfer is completed. It can reach a large air flux. The quick sampling connector is composed of a sampling nozzle and a one-way valve, which can prevent the liquid from flowing backwards during the sampling process and ensure the aseptic sampling.
- Ø:—
- Désignation:Sterile PETG square storage bottle bi-directional transfer cap with ¹/₄" TPE tube, female Luer connector with male sealing plug vent filter
- Longueur:—
- For Use With:2 L Flasks