Colored Cap Inserts
Coloured cap insert, Blue
Catalog # 77508-332
Fournisseur #: 611004
Jump to:
Discontinued
Product Details & Documents
Six colored cap inserts for easy identification. Compatible with regular cryogenic vials that with external thread caps only.
Packaging: 100/pack; 1000/case
Spécifications
- Color:Blue
- Désignation:Coloured cap insert