Colored Cap Inserts
Coloured cap insert, Red
Catalog # 77508-328
Supplier: NEST SCIENTIFIC
Supplier #: 611002
Jump to:
Product Details & Documents
Six colored cap inserts for easy identification. Compatible with regular cryogenic vials that with external thread caps only.
Packaging: 100/pack; 1000/case
Spécifications
- Color:Red
- Désignation:Coloured cap insert