Deep Well Plates
Product Details & Documents
The deep well plates have a variety of specifications, conforming to the needs of fully automatic workstations and experiments.
- High quality imported PP material employed for high stability and no chemical reactions with test reagents
- Maximum sustainable centrifuge force 4000×g
- Compatible with DMSO and inert to water
- Can be stored under subzero temperature from −40 to −80 ℃
- Autoclavable at 20 psi, 121 ℃ for 20 minutes, great heating uniformity
- Minimum residual liquid, low heavy metal content
Genomic DNA extraction, plasmid DNA extraction, and nucleic acid extraction and purification of various samples. High-throughput automatic liquid operation to achieve high-throughput operations, such as protein precipitation, liquid extraction, and processing of animal tissues, bacteria, plants, soil, clinical samples, yeasts and more.
These plates have flat bottom and can be transported for long distance if sealed by heat sealable film. These are produced and packed in Class 100000 clean workshop.
They have alphabetical sorting and corner cut marking for convenient tracking of samples. Each package has a separate article number/batch number identification, facilitating the quality tracking.
Certifications: Certified DNase/RNase and pyrogen free. Conformed to international SBS standards.
Spécifications
- Sterility:Non sterile
- Volume:0.4 ml
- Désignation:Deep well plate, 96-well
- Packaging:5/pack, 50/case
- Well Bottom Shape:U-bottom



