Transfer Caps for Storage Bottles, Square, Closed System Solution
Transfer Caps for Storage Bottles, Square, Closed System Solution
Bi-directional transfer cap, 500 ml Square storage bottle closed system solution
Catalog # 77509-148
Supplier:   NEST SCIENTIFIC
Supplier #:  C51021-BHB060B
Transfer Caps for Storage Bottles, Square, Closed System Solution
Catalog # 77509-148
Supplier:  NEST SCIENTIFIC
Transfer Caps for Storage Bottles, Square, Closed System Solution
Bi-directional transfer cap, 500 ml Square storage bottle closed system solution
Price
1 946,11 $
Log in pour la disponibilité
Case of 10 Each
Product Details & Documents
These injection-molded transfer caps are highly compatible with other major brands.
Spécifications
  • Désignation:
    Bi-directional transfer cap
  • For Use With:
    500 ml Square storage bottle closed system solution