Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Erlenmeyer Flasks, PETG, Flat Bottom

Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per case, 125 ml, Sterile, 106.8 mm, 34 mm
  77508-806
 :  781011
Erlenmeyer Flasks, PETG, Flat Bottom
  77508-806
Erlenmeyer Flasks, PETG, Flat Bottom

Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per case, 125 ml, Sterile, 106.8 mm, 34 mm

 
$209.04
   
Case of 24 Each
 

Ideal for shaker culture applications. PETG Erlenmeyer flasks are manufactured in 100000 grade cleaning room with USP 6 grade raw materials.

  • PETG Bottle and HDPE cap
  • Sterilized by E-Beam
  • Nonpyrogenic
  • Packaged in sterile bags

Vent caps with 0.22 µm hydrophobic filters for gas exchange without contamination.

 : ISO 9001 and 13485 certified.

 
  • Sterility:
    Sterile
  • Capacité:
    125 ml
  • Désignation:
    Conical PETG Erlenmeyer flasks, flat bottom, Bottom Ø: 66 mm, 24 per case
  • Hauteur:
    106.8 mm
  • Neck O.D.:
    34 mm