Universal Pipette Tips | Avantor
Universal Pipette Tips
Graduated, extra long, 10 µl, -, Clear, 96/rack, 960/pack, 9600 case
Catalog # 77507-500
Supplier:   NEST SCIENTIFIC
Fournisseur #:  304023
Universal Pipette Tips
Catalog # 77507-500
Supplier:  NEST SCIENTIFIC
Universal Pipette Tips
Graduated, extra long, 10 µl, -, Clear, 96/rack, 960/pack, 9600 case
Price
$528.12
Log in pour la disponibilité
Case of 9,600 Each
Product Details & Documents
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
  • Universal fit: Suitable for use with most pipettors
  • Withstands organic solvents
  • Less residue
  • Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
 : Available in different packaging options: Bulk, racked, stack racked or in refills.
Spécifications
  • Volume:
    10 µl
  • Tip Color:
    Clear
  • Packed:
    96/rack, 960/pack, 9600 case
  • Sterile:
    -
  • Tip Style:
    Graduated, extra long