Universal Pipette Tips
Graduated, extra long, 10 µl, +, Clear, 96/box, 4800/case
Catalog # 77507-498
Supplier: NEST SCIENTIFIC
: 304012
Jump to:
Product Details & Documents
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
Universal fit: Suitable for use with most pipettors
- Withstands organic solvents
- Less residue
- Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
: Available in different packaging options: Bulk, racked, stack racked or in refills.
Spécifications
- Volume:10 µl
- Tip Color:Clear
- Packed:96/box, 4800/case
- Sterile:+
- Tip Style:Graduated, extra long