Universal Pipette Tips
Graduated, extra long, 10 µl, -, Clear, 1000/pack, 10000/case
  77507-496
 :  304001
Universal Pipette Tips
  77507-496
Universal Pipette Tips
Graduated, extra long, 10 µl, -, Clear, 1000/pack, 10000/case
$294.51
Case of 10,000 Each
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
  • Universal fit: Suitable for use with most pipettors
  • Withstands organic solvents
  • Less residue
  • Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
 : Available in different packaging options: Bulk, racked, stack racked or in refills.
  • Volume:
    10 µl
  • Tip Color:
    Clear
  • Packed:
    1000/pack, 10000/case
  • Sterile:
    -
  • Tip Style:
    Graduated, extra long