Universal Pipette Tips
Graduated, extra long, 10 µl, -, Clear, 1000/pack, 10000/case
77507-496
: 304001
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
Universal fit: Suitable for use with most pipettors
- Withstands organic solvents
- Less residue
- Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
: Available in different packaging options: Bulk, racked, stack racked or in refills.
- Volume:10 µl
- Tip Color:Clear
- Packed:1000/pack, 10000/case
- Sterile:-
- Tip Style:Graduated, extra long