Storage Bottles, Round
Product Details & Documents
Round Storage Bottles are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
- Excellent physical and chemical indicators
- Good pressure-resistance
- Impact-resistance
- Acid and alkali resistance
- Autoclave sterilization at 121 ℃, HDPE can be stored at −80 ℃
- No biotoxicity, pyrogen-free, DNAse/RNAse-free
Sterile and enzyme-free plastic reagent bottles, suitable for product packaging and storage requirements in molecular biology and Cell Biology, laboratory medicine, genomics and proteomics and other areas. Improvement of the quality of product packaging helps to improve the company image and brand value, lets your product packaging meet international standards and enter the international market.
Washing-free, no need for tedious pre-cleaning processing work, ready to use, greatly improves the packaging effect of users. Thickened middle packaging to ensure transportation and storage safety. Use of professional leaking-proof design for bottle mouth, with no need for inner cover or inner gasket for protection, excellent sealability. Guarantee of being 100% leaking-proof to ensure safety even during air transportation, wide-mouthed design for easy access of liquid.
Certifications: ISO 9001&13485 certified.
Spécifications
- Volume:15 ml
- Color:Amber
- Matière:HDPE
- Packaging:20/pack, 400/case
- Bottle Style:Round
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



