Inoculating Needle/Loops
Inoculating needle, White, 400/pack, 4000/case
Catalog # 77508-606
Supplier: NEST SCIENTIFIC
Fournisseur #: 716001
Jump to:
Product Details & Documents
Inoculating needle/loops made up of PP material, hydrophilic.
- Applied in microbiology experiments
- Different colors for identification
- Sterilized by ethylene oxide
- Easy-tear sterile individual packaging
Flexible handles for easier collection and inoculation.
Spécifications
- Color:White
- Désignation:Inoculating needle
- Packaging:400/pack, 4000/case