Robotic Pipette Tips | Avantor
Robotic Pipette Tips
1000 µl, +, 96 pieces/box, 10 boxes/pack, 5 packs/case
Catalog # 77507-880
Supplier:   NEST SCIENTIFIC
Fournisseur #:  345719
Robotic Pipette Tips
Catalog # 77507-880
Supplier:  NEST SCIENTIFIC
Robotic Pipette Tips
1000 µl, +, 96 pieces/box, 10 boxes/pack, 5 packs/case
Price
841,41 $
Log in pour la disponibilité
Case of 4,800 Each
Product Details & Documents
NEST robotic conductive tip can be applied in R and D of genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals and other high flux liquid transfer.
  • Made of polymer material polypropylene (PP)
  • Offers high efficiency and repeatability
  • With barcode
Surface owns high hydrophobicity as treated with a special technique making for certain purposes.
Spécifications
  • Volume:
    1000 µl
  • Packed:
    96 pieces/box, 10 boxes/pack, 5 packs/case
  • Sterile:
    +