Robotic Pipette Tips
1000 µl, +, 96 pieces/rack 24 racks/pack
Catalog # 77507-874
Supplier: NEST SCIENTIFIC
Fournisseur #: 345703
Jump to:
Product Details & Documents
NEST robotic conductive tip can be applied in R and D of genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals and other high flux liquid transfer.
- Made of polymer material polypropylene (PP)
- Offers high efficiency and repeatability
- With barcode
Surface owns high hydrophobicity as treated with a special technique making for certain purposes.
Spécifications
- Volume:1000 µl
- Packed:96 pieces/rack 24 racks/pack
- Sterile:+