Universal Pipette Tips
Graduated, 200 µl, -, Jaune, 1000/pack, 20000/case
  77507-484
 :  302106
Universal Pipette Tips
  77507-484
Universal Pipette Tips
Graduated, 200 µl, -, Jaune, 1000/pack, 20000/case
515,79 $
Case of 20,000 Each
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
  • Universal fit: Suitable for use with most pipettors
  • Withstands organic solvents
  • Less residue
  • Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
 : Available in different packaging options: Bulk, racked, stack racked or in refills.
  • Volume:
    200 µl
  • Tip Color:
    Jaune
  • Packed:
    1000/pack, 20000/case
  • Sterile:
    -
  • Tip Style:
    Graduated