Universal Pipette Tips
Graduated, 200 µl, -, Jaune, 1000/pack, 20000/case
77507-484
: 302106
Nest Scientific Universal Fit Pipette Tips are made from premium-grade 100% polypropylene and engineered for the most demanding molecular biology applications.
Universal fit: Suitable for use with most pipettors
- Withstands organic solvents
- Less residue
- Made from low retention plastic
Sterilized by gamma irradiation (only valid for some variants), they are certified RNase-, DNase-, human DNA-, and pyrogen-free.
: Available in different packaging options: Bulk, racked, stack racked or in refills.
- Volume:200 µl
- Tip Color:Jaune
- Packed:1000/pack, 20000/case
- Sterile:-
- Tip Style:Graduated