Serological Pipettes
1/pack, 50/box, 300/case
Catalog # 77507-640
Supplier: NEST SCIENTIFIC
Fournisseur #: 329501
Jump to:
Product Details & Documents
Serological pipettes can be used for liquid transfer in biological experiments such as cell culture and bacterial culture.
- Made of high clarity polystyrene
- Sterilized by E-beam
- Non-pyrogenic, non-cytotoxic and non-hemolytic
- DNase/RNase free
- Polyolefin fiber filter to reduce contamination
- Edged color-coded band for identification
- Negative graduations for extra volume
Documents
Spécifications
- Packed:1/pack, 50/box, 300/case