AlphaMop™ Polyester Mop | Avantor
AlphaMop™ Polyester Mop
AlphaMop™ polyester replacement covers and pads: STX7118, for use with TX7108 and TX7108A, sterile, 38.1×20.32 cm, 10 covers and 1 pad/bag; 12 bags/case
Catalog # TWSTX7118
Supplier:   Texwipe
Fournisseur #:  STX7118
AlphaMop™ Polyester Mop
Catalog # TWSTX7118
Supplier:  Texwipe
AlphaMop™ Polyester Mop
AlphaMop™ Polyester Mop
AlphaMop™ polyester replacement covers and pads: STX7118, for use with TX7108 and TX7108A, sterile, 38.1×20.32 cm, 10 covers and 1 pad/bag; 12 bags/case
Price
$1,193.10
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 120
Product Details & Documents
Ideal for cleaning walls, floors, and other flat surfaces in critical environments.
AlphaMop™ covers TWTX7026 are pre-wetted with 6% isopropyl alcohol. The case purchase includes two foam pads. Replacement pack TW-TX7118 contains 150 polyester heads and 6 foam pads.
The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fiberglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. Lightweight mop maintains ultralow generation of particles and extractables. Bag-within-a-Bag® packaged.
Spécifications
  • Désignation:
    AlphaMop™ polyester replacement covers and pads: STX7118, for use with TX7108 and TX7108A, sterile
  • L×l:
    38.1×20.32 cm
  • Packed:
    10 covers and 1 pad/bag; 12 bags/case