Hydroxylammonium chlorure solution 100 g/l
Catalog # 104086-918
Marketsource
Supplier #: 81842
Hydroxylammonium chlorure solution 100 g/l
Catalog # 104086-918
Marketsource
CAS Number: 5470-11-1
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
HYDROXYLAMINE HYDROCHLOR 16OZ Catalog #RC390016
Product RC390016 is a replacement for 104086-918 and has the same packaging and specifications, but may be a different quantity.
HYDROXYLAMINE HYDROCHL SOLN Catalog #CA11016-776
Product CA11016-776 is a replacement for 104086-918 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
For determination of volatile acids by the esterification method
Hach method 8196 (100 tests).
Synonymes: Hydroxylamine chlorhydrate
- CAS Number:5470-11-1
- Formule:NH₂OH·HCl
- Numéro MDL:MFCD00051089
- Molecular Weight:69.49 g/mol
- Température de conservation:Température ambiante
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



