Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Adjustable Wrist Grounding Strap, STATICO | Avantor

Adjustable Wrist Grounding Strap, STATICO

Wrist strap with coil cord and alligator clip, 1.8 m (6')
Catalog # 14223-709
Supplier:   Statico
Fournisseur #:  S0101
Adjustable Wrist Grounding Strap, STATICO
Catalog # 14223-709
Supplier:  Statico
Adjustable Wrist Grounding Strap, STATICO
Adjustable Wrist Grounding Strap, STATICO

Wrist strap with coil cord and alligator clip, 1.8 m (6')

Price
28,84 $
Log in for availability
Each
Product Details & Documents

Adjustable light blue wrist grounding strap with 360⁰ skin contact

A 1 Meg current limiting resistor is molded-in at the snap end termination of the cord

Certifications: Meets ANSI/ESD S1.1-2013 & ANSI/ESD S20.20-2014.

Ordering information: Available with either 1.8 m (6') or 3.6 m (12') Coil Cord and an Alligator Clip.

Specifications
  • Désignation:
    Wrist strap with coil cord and alligator clip
  • Cord Length:
    1.8 m (6')
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.