ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 10 ml
Catalog # 77461-918
Supplier: AAT Bioquest
Fournisseur #: 22870
ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
Catalog # 77461-918
Supplier: AAT Bioquest
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The ReadiUse™ Annexin Binding Buffer *Optimized for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.
- Provided as a 10X concentrate
- Saves time
- pH of 7.4
Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.
: For research use only.
Spécifications
Spécifications
- Désignation:ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 10 ml
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



