Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes | Avantor

NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes

NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (193-288), 192
Catalog # CA77448-124
Fournisseur #:  NOVA-512922
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
Catalog # CA77448-124
CAS Number:  
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes
NEXTFLEX™ RNA-Sequence 2.0 UDI Barcodes

NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (193-288), 192

CAS Number:  
Price
$1,384.31
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

The NEXTFLEX™ RNA-sequence 2.0 UDI barcodes are designed to prepare multiplexed single and paired-end cDNA libraries from total RNA, mRNA-enriched RNA or rRNA-depleted RNA for Illumina platforms.

  • UDI barcodes for multiplexing RNA-sequence libraries
  • Compatible with both paired-end and single-read sequencing
  • Every lot is tested for index purity and functionality by sequencing

The NEXTFLEX™ unique dual index barcodes contain two unique indexes on each adapter, with a hamming distance of at least three throughout the entire series, to allow error correction. The NEXTFLEX™ unique dual index RNA-sequence barcodes can be used for PCR-free RNA sequencing applications. Patterned flow cells found on the Illumina® Novasequence®, Hisequence® 3000/4000, and Hisequence® X platforms suffer from increased rates of sample mis-assignment during sequencing. The use of adapters with unique dual indices during sequencing prevents such mis-assigned reads from appearing in final data sets, allowing for the highest assurance of data integrity. Every lot is tested for index purity and functionality by sequencing.

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Spécifications

Spécifications

  • Désignation:
    NEXTFLEX™ RNA-sequence 2.0 UDI barcodes (193-288)
  • No. of Reactions:
    192

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.