BioXp® Tip Kits | Avantor
BioXp® Tip Kits
5×50 µl rack, 3×200 µl rack
Catalog # 77461-214
Supplier:   TELESIS BIO
Fournisseur #:  12214KT
BioXp® Tip Kits
Catalog # 77461-214
Supplier:  TELESIS BIO
BioXp® Tip Kits
BioXp® Tip Kits
5×50 µl rack, 3×200 µl rack
Price
$193.67
Log in pour la disponibilité
Pack of 1
La quantité minimale de commande est de 1
Product Details & Documents
BioXp® system tips in various kit types and sizes.
  • Free of human DNA
  • Rnase/Dnase free
  • PCR inhibitor free
  • Automated
Different kit types and sizes will require different quantities of 50 and 200 µl racks.
Ordering information: Verify the tip requirements for the BioXp® kit that you are ordering.
Spécifications
  • Désignation:
    5×50 µl rack, 3×200 µl rack