Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
NEXTFLEX™ Poly(A) Beads 2.0 | Avantor

NEXTFLEX™ Poly(A) Beads 2.0

NEXTFLEX™ Poly(A) beads 2.0, 8
Catalog # CA77448-138
Fournisseur #:  NOVA-512991
NEXTFLEX™ Poly(A) Beads 2.0
Catalog # CA77448-138
CAS Number:  
NEXTFLEX™ Poly(A) Beads 2.0
NEXTFLEX™ Poly(A) Beads 2.0

NEXTFLEX™ Poly(A) beads 2.0, 8

CAS Number:  
Price
$64.13
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

NEXTFLEX™ poly(A) beads 2.0 kits offer a convenient method for obtaining pure, intact mRNA upstream of RNA-sequence library prep with improved mRNA yields, low rRNA contamination and a simple protocol.

  • 10 ng - 5 µg total starting RNA
  • Magnetic bead-based protocol
  • No organic solvents or precipitation step required
  • Optimized for use with 10 ng - 5 µg of total RNA as starting material
  • Versions are available for automated and manual isolation of the mRNA transcriptome
  • Automated on the Sciclone® G3 NGSx and Zephyr® G3 NGS workstations

Several approaches have been used to remove ribosomal RNA (rRNA) from total RNA samples for RNA-sequence library preparation. Removal of rRNA avoids the waste of reagents and bioinformatics resources to sequence and align ~85% of total RNA comprising rRNA (which is generally not a target of interest in NGS sequencing experiments). Revvity’s NEXTFLEX™ poly(A) beads 2.0 kit offers an easy and cost-effective method for removing rRNA in RNA-sequence libraries. This product takes advantage of the tract of adenosine residues present at the 3’ ends of a vast majority of protein-coding mRNAs. Hybridization of these poly(A) tails to tracts of complementary thymidines ('oligo dT') immobilized on solid supports allows the retrieval of poly(A)+ RNAs by recovering the solid support along with the hybridized RNA complexes.

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Spécifications

Spécifications

  • Désignation:
    NEXTFLEX™ Poly(A) beads 2.0
  • No. of Reactions:
    8

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.