Magleads
Magleads, pack of 10, Black, 12"
  470355-432
 :  ML300B
Magleads
  470355-432
Magleads
Magleads
Magleads, pack of 10, Black, 12"
$39.82
Pack of 10
Simply the easiest and best jumper wires you can have in your classroom.
  • The 4 mm diameter magnets work in all the places that standard clips work
  • No need for an adapter to connect wires to your battery
  • No more fighting with Fahnestock or alligator clips - just click them together!
Instantly make working electrical circuits with these ingenious leads, each fitted with a tiny powerful magnet at either end. Place a couple near to a battery case and they connect by themselves. Then, place them next to a bulb holder connector and you have your first working circuit!
  • Color:
    Black
  • Désignation:
    Magleads, pack of 10
  • Longueur:
    12"