BioXp® Tip Kits
3×50 µl rack, 1×200 µl rack
Catalog # 77461-184
Supplier: TELESIS BIO
: 12025KT
Jump to:
Product Details & Documents
BioXp® system tips in various kit types and sizes.
- Free of human DNA
- Rnase/Dnase free
- PCR inhibitor free
- Automated
Different kit types and sizes will require different quantities of 50 and 200 µl racks.
: Verify the tip requirements for the BioXp® kit that you are ordering.
Spécifications
- Désignation:3×50 µl rack, 1×200 µl rack