Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Magleads

Magleads, pack of 10, Red, 12"
Catalog # 470355-440
Supplier:   American Scientific
 :  ML300R
Magleads
Catalog # 470355-440
Magleads
Magleads

Magleads, pack of 10, Red, 12"

Price
$39.82
Log in for availability
Pack of 10
Product Details & Documents

Simply the easiest and best jumper wires you can have in your classroom.

  • The 4 mm diameter magnets work in all the places that standard clips work
  • No need for an adapter to connect wires to your battery
  • No more fighting with Fahnestock or alligator clips - just click them together!

Instantly make working electrical circuits with these ingenious leads, each fitted with a tiny powerful magnet at either end. Place a couple near to a battery case and they connect by themselves. Then, place them next to a bulb holder connector and you have your first working circuit!

Specifications
  • Color:
    Red
  • Désignation:
    Magleads, pack of 10
  • Longueur:
    12"