Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits | Avantor

NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits

NEXTFLEX™ Rapid XP V2 DNA-sequence kit, 8
Catalog # CA77448-180
Fournisseur #:  NOVA-5149-21
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
Catalog # CA77448-180
CAS Number:  
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits
NEXTFLEX™ Rapid XP V2 DNA-Sequence Kits

NEXTFLEX™ Rapid XP V2 DNA-sequence kit, 8

CAS Number:  
Price
$302.53
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

The NEXTFLEX® Rapid XP V2 DNA-sequence kit offers a simplified DNA-seq library prep workflow for Illumina® platforms. Libraries produced using this kit are normalized during library construction.

  • Fully automated normalization and fragmentation
  • Enzymatic fragmentation with end-repair and A-tailing in one reaction
  • Highly efficient ligation with low adapter dimer formation
  • Efficient: Automated on the Sciclone®  and Zephyr®  NGS workstations for mid-high throughput automation and now on the BioQule™ NGS System for low throughput labs

The NEXTFLEX® Rapid XP V2 DNA-seq kit includes proprietary NEXTFLEX® normalization beads, which provide a consistent mass and sequencing cluster density for all samples in a library pool. This shortens the time needed for quantification and pooling preparation for sequencing in a high throughput lab by up to 3 hours per 96 samples. The NEXTFLEX  Rapid XP V2 DNA-seq kit offers a streamlined workflow with highly efficient ligation, generating low amount of adapter dimers even at low input concentrations.

The NEXTFLEX Rapid XP V2 DNA-Seq kit is compatible with multiple liquid handlers, including the Sciclone G3 NGSx workstation, Sciclone G3 NGSx iQ™ workstation, and Zephyr NGS workstation. For low throughput automation, the NEXTFLEX  Rapid XP V2 DNA-Seq kit is also now automated on the BioQule™ NGS System which simplifies both library prep and quantitation. This low-cost, benchtop instrument delivers libraries ready to load into your sequencer with only 15 minutes of hands-on time. 

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Spécifications

Spécifications

  • Désignation:
    NEXTFLEX™ Rapid XP V2 DNA-sequence kit
  • No. of Reactions:
    8

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.