Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
NEXTFLEX™ Universal Blockers | Avantor

NEXTFLEX™ Universal Blockers

NEXTFLEX™ universal blockers, 96
Catalog # CA77448-178
Fournisseur #:  NOVA-5143233
NEXTFLEX™ Universal Blockers
Catalog # CA77448-178
CAS Number:  
NEXTFLEX™ Universal Blockers
NEXTFLEX™ Universal Blockers

NEXTFLEX™ universal blockers, 96

CAS Number:  
Price
$497.35
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

The NEXTFLEX™ universal blockers block non-specific hybridization, enhancing specificity and reducing sequencing costs. They are compatible with all ligation and tagmentation workflows for Illumina®

  • Effective in blocking single-index or dual-index libraries
  • Compatible with all NEXTFLEX™ barcodes and all other ligation-based and tagmentation-based workflows for Illumina® sequencing platforms
  • Simplifies and reduces the cost of target capture without sacrificing performance
  • 96 barcode blockers available

During target enrichment, adapters can interact with the complementary adapter sequence strand of another library molecule. The NEXTFLEX™ universal blockers are designed to prevent this concatemerization of library molecules. By blocking this complementary adapter sequence interaction, the number of on-target reads improves dramatically and the depth of enrichment increases. The NEXTFLEX™ universal blockers are designed for use with indexed adapters for Illumina sequencing platforms. They are effective in blocking single-indexed or dual-indexed libraries, without being affected by index length or the presence of a unique molecular index (UMI). The NEXTFLEX Universal Blockers are compatible with ligation-based workflows, such as NEXTFLEX™ or Illumina trusequence library prep kits. They are also compatible with tagmentation-based workflows, such as Illumina Nextera library prep kits.

 : Manufactured in ISO 9001 facility.

 : For research use only. Not for use in diagnostic procedures.

Spécifications

Spécifications

  • Désignation:
    NEXTFLEX™ universal blockers
  • No. of Reactions:
    96

Spécifications