STREPTAVIDIN HOOK MALEIMIDE ACT 1MG
Catalog # CA77448-434
Supplier: G-Biosciences
STREPTAVIDIN HOOK MALEIMIDE ACT 1MG
Catalog # CA77448-434
Supplier: G-Biosciences
Supplier Number: 786-1653
Frequently Bought Together
Acetonitrile ≥99.8% for HPLC, Supelco®
Supplier: MilliporeSigma
Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Supplier: VWR International
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
VWR® Sleeve Stoppers
Supplier: VWR International
TETRAHYDROFURAN OMNISOLV 4L
Supplier: MilliporeSigma
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
SingleQuots® EGM®-2 Endothelial Kit
Supplier: LONZA PHARMA - BIOSCIENCE CA
Toluène ≥99.5% (by GC), AR® (ACS) ACS analytical reagent, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Glycine ≥98.5% ACS
Supplier: Thermo Scientific Chemicals
E. cloni® Electrocompetent Cells, Biosearch Technologies
Supplier: Lucigen
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
Tetrahydrofuran, anhydrous ≥99.8%, AcroSeal™ BHT-free, over molecular sieves
Supplier: Thermo Scientific Chemicals
Griffin Style Beakers, Mortech
Supplier: Mortech Manufacturing