Epic High Performance Lab Coats
Blue, 7XL
Catalog # 77448-280
Supplier: TIANS INTERNATIONAL INC. TE
: 8647956-7XL
Jump to:
Product Details & Documents
Low-linting lab coats for use in critical and low particulate cleanroom environments.
Low-linting, high filtration fabric
- Excellent splash protection
- Snap front, knit wrist, knit collar
- Three pockets
- Helmke Drum tested
Fabric is constructed from a three-layer, spun-bonded material that contains a microscopic filtration layer. Individually inspected for sizing, stitching, and workmanship. High filtration fabric holds out 98.8% of particles at 3 µm, and also provides excellent splash protection from non-hazardous fluids. Features include a snap front, knit wrists, knit collar, and three pockets.
: ISO class 6.
: Neatly folded and cleanroom packed. 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case.
Spécifications
- Taille de confection:7XL
- Color:Blue
- Cat. No.:77448-280
- Cleanroom class:1000/ISO 6
- Garment application:Cleanroom
- Pockets:With pockets
- Closure type:Press studs / snaps
- Weight:60 gm/m²



