CARTRIDGE FILTER POLYFLOW-G 5UM PP 3 MM
Catalog # 77441-822
Supplier: PARKER HANNIFIN TE
CARTRIDGE FILTER POLYFLOW-G 5UM PP 3 MM
Catalog # 77441-822
Supplier: PARKER HANNIFIN TE
Supplier Number: PG-10010-050-14
Frequently Bought Together
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Single-Use Needles, BD Medical
Supplier: BD
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Calibrated Loops, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Ammonia solution 28.0 - 30.0% (NH₃ basis), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Corning® Cell Recovery Solution, Corning
Supplier: Corning
CL17 DPD Indicator, Hach
Supplier: Hach