FILTER CARTRIDGE PROFLOWII-G EPDM
77441-802
Frequently Bought Together
TETRAHYDROFURAN OMNISOLV 4L
: MilliporeSigma
Phenol 99+% stabilized ACS
: Thermo Scientific Chemicals
2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC
SingleQuots® EGM®-2 Endothelial Kit
: LONZA PHARMA - BIOSCIENCE CA
Square Petri Dish, Electron Microscopy Sciences
: Electron Microscopy Sciences
Glycine ≥98.5% ACS
: Thermo Scientific Chemicals
TrueBlot® IP/Western Blot, Rockland Immunochemicals
: Rockland Immunochemical
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC