CARTRIDGE CLEARFLOW B 0.2UM O-RING
Catalog # 77441-850
Supplier: PARKER HANNIFIN TE
CARTRIDGE CLEARFLOW B 0.2UM O-RING
Catalog # 77441-850
Supplier: PARKER HANNIFIN TE
Supplier Number: SG-60720-002-2-B
Frequently Bought Together
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
Self Standing Sample Tubes, 5 and 10 ml, Globe Scientific
Supplier: Globe Scientific
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Sodium hydroxyde 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
STERI-STRIP CLOSURES 3MMX7.6CM
Supplier: Safecross First Aid Limited
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
LONGAMP® Hot Start Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
EDTA disodium salt dihydrate 99+%
Supplier: Thermo Scientific Chemicals
1,2-Bis(diphenylphosphino)ethane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
SingleQuots® EGM®-2 Endothelial Kit
Supplier: LONZA PHARMA - BIOSCIENCE CA
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
Ethyl methanesulfonate ≥99.0% (by GC)
Supplier: TCI America
E. cloni® Electrocompetent Cells, Biosearch Technologies
Supplier: Lucigen
CitriSolv™ Xylene Substitute, Hybrid
Supplier: Decon Labs
Standard Pin Stub SEM Mount Slotted Head, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences