Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Duplex Dnase

Duplex Dnase

Duplex DNase, 1000 units
Catalog # 77441-306
Supplier #:  M7635S
Duplex Dnase
Catalog # 77441-306
CAS Number:  
Duplex Dnase

Duplex DNase, 1000 units

CAS Number:  
Price
261,65 $
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

An engineered double-strand specific DNA endonuclease, dsDNase preferentially degrades double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA) or RNA.

  • Specifically degrades dsDNA in the presence of ssDNA
  • Cleaves DNA strand of DNA:RNA hybrid duplex
  • Isolated from a recombinant sourceRNase-free
  • Supplied with 10X reaction buffer

This specificity toward dsDNA allows the enzyme to degrade contaminating dsDNA while maintaining the integrity of single stranded DNA molecules such as primers and probes within PCR mixes, allowing for efficient decontamination of PCR master mixes. Moreover, dsDNase displays the unique property of cleaving the DNA strand of a DNA:RNA hybrid duplex.

Concentration: 2000 units/ml

Specifications

Specifications

  • Désignation:
    Duplex DNase
  • Taille:
    1000 units

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.