Compresstome® Agarose tablets
Compresstome® Agarose tablets
Compresstome® Agarose tablets
Catalog # 77435-392
Supplier #:  VFAGTVM20CANAL
Compresstome® Agarose tablets
Catalog # 77435-392
Compresstome® Agarose tablets
Compresstome® Agarose tablets
Compresstome® Agarose tablets
Price
$126.75
Log in for availability
Pack of 20
Product Details & Documents
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
  • Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
Specifications
  • Désignation:
    Compresstome® Agarose tablets