Compresstome® Agarose tablets
Compresstome® Agarose tablets
Catalog # 77435-390
Supplier: PRECISIONARY INSTRUMENTS LLC
Fournisseur #: VFAGTVM10CANAL
Jump to:
Product Details & Documents
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
- Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
Spécifications
- Désignation:Compresstome® Agarose tablets