Compresstome® Agarose tablets | Avantor
Compresstome® Agarose tablets
Compresstome® Agarose tablets
Catalog # 77435-390
Fournisseur #:  VFAGTVM10CANAL
Compresstome® Agarose tablets
Catalog # 77435-390
Compresstome® Agarose tablets
Compresstome® Agarose tablets
Compresstome® Agarose tablets
Price
$66.29
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
  • Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
Spécifications
  • Désignation:
    Compresstome® Agarose tablets