Compresstome® Agarose tablets
Compresstome® Agarose tablets
  77435-388
 :  VFAGT100CANAL
Compresstome® Agarose tablets
  77435-388
Compresstome® Agarose tablets
Compresstome® Agarose tablets
Compresstome® Agarose tablets
544,03 $
Pack of 100
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
  • Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
  • Désignation:
    Compresstome® Agarose tablets