Compresstome® Agarose tablets
Compresstome® Agarose tablets
77435-388
: VFAGT100CANAL
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
- Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
- Désignation:Compresstome® Agarose tablets