Compresstome® Training Plan Service
Compresstome training demo basic plan
  77435-500
 :  VFTDBP
Compresstome® Training Plan Service
  77435-500
Compresstome® Training Plan Service
Compresstome training demo basic plan
$871.82
Each
Service plans for either a basic or premium compresstome training after a researcher receives the machine.
  • Premium or basic compresstome training demos
Premium plan for training on the compresstome features live video session with compresstome technician on using the tissue slicer, with explanation of main compresstome slicer functions, embedding tissue, making agarose solution, sectioning sample tissue, and cleaning and maintenance of the compresstome, including time with chief scientific officer.
Basic plan for training on the compresstome features video session with compresstome technician on using the tissue slicer from beginning to end, including explanation of main compresstome slicer functions, embedding tissue, making agarose solution, sectioning sample tissue, and cleaning and maintenance.
  • Désignation:
    Compresstome training demo basic plan