Compresstome® Sectioning Tools | Avantor
Compresstome® Sectioning Tools
Fine tissue handling brushes
Catalog # 77435-532
Fournisseur #:  VFVMPBCANAL
Compresstome® Sectioning Tools
Catalog # 77435-532
Compresstome® Sectioning Tools
Compresstome® Sectioning Tools
Fine tissue handling brushes
Price
$118.63
Log in pour la disponibilité
Pack of 3
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sectioning tools designed for use with compresstome rotary microtomes.
  • Fine tissue forceps are supplied individually
  • Tissue paintbrushes are supplied in packs of 3
Fine tissue handling forceps enable gentle manipulation of tissues and organs during dissection. They allow for the gentle removal of the tissue from the bufffer tray while slicing.
Tissue paintbrushes allow for easy handling of delicate tissue slices. Soft bristles help prevent damage to tissue sections when transferring slices.
Spécifications
  • Désignation:
    Fine tissue handling brushes