Compresstome® Sectioning Tools
Compresstome® Sectioning Tools
Fine tissue handling forceps
  77435-462
Supplier #:  VFFORCEPS
Compresstome® Sectioning Tools
  77435-462
Compresstome® Sectioning Tools
Compresstome® Sectioning Tools
Fine tissue handling forceps
$28.17
Each
Sectioning tools designed for use with compresstome rotary microtomes.
  • Fine tissue forceps are supplied individually
  • Tissue paintbrushes are supplied in packs of 3
Fine tissue handling forceps enable gentle manipulation of tissues and organs during dissection. They allow for the gentle removal of the tissue from the bufffer tray while slicing.
Tissue paintbrushes allow for easy handling of delicate tissue slices. Soft bristles help prevent damage to tissue sections when transferring slices.
  • Désignation:
    Fine tissue handling forceps