Wallcur® PRACTI-Labeled Vials | Avantor
Wallcur® PRACTI-Labeled Vials
Practi-cefoxitin (2 g/ 20 ml), Unsterile fructose powder
Catalog # 470355-018
Supplier:   WALLCUR, LLC.
Fournisseur #:  493CF
Wallcur® PRACTI-Labeled Vials
Catalog # 470355-018
Supplier:  WALLCUR, LLC.
Wallcur® PRACTI-Labeled Vials
Wallcur® PRACTI-Labeled Vials
Practi-cefoxitin (2 g/ 20 ml), Unsterile fructose powder
Price
$172.22
Log in pour la disponibilité
Box of 30
Product Details & Documents
Prelabeled Vials For Convenience
  • Contains No Medication
  • Suitable For Most Simulator Applications
Each of these vials is already labeled in the same way as their real counterparts are! Available in a wide variety of simulated labels, each contains all the information a normal label does as well as additional warnings for training purposes. Vials are also bar coded making your pharma training easier for inventory management. Materials containing only Distilled water are generally suitable for simulator use. Those containing other materials are not recommended for use in simulators. All of these factors make these vials ideal for skills training for nursing, EMS and pharm tech programs.
Have students practice all the skills needed for proper drug selection, dosing and administration. These Vials help teach:
Reading and interpreting dosage requirements
Calculating commonly ordered antihistamine drug dosages
Small vial pressure changes
Small needle syringe techniques
Injection administration skills
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Spécifications
  • Désignation:
    Practi-cefoxitin (2 g/ 20 ml)
  • Includes:
    Unsterile fructose powder