Wallcur® PRACTI-Rapid Sequence Intubation Training Kit
Wallcur® PRACTI-Rapid Sequence Intubation Training Kit
Practi-RSI refill kit
Catalog # 470354-944
Supplier:   WALLCUR, LLC.
Supplier #:  705RF
Wallcur® PRACTI-Rapid Sequence Intubation Training Kit
Catalog # 470354-944
Supplier:  WALLCUR, LLC.
Wallcur® PRACTI-Rapid Sequence Intubation Training Kit
Wallcur® PRACTI-Rapid Sequence Intubation Training Kit
Practi-RSI refill kit
Price
$110.21
Log in for availability
Kit of 1
Product Details & Documents
By Educator Request!
  • Contains No Medication
  • Contains No Needles
We now offer the EMS/Paramedic Rapid Sequence Intubation (RSI) Kit for clinical training from Wallcur without additional syringes or needles. The PRACTI-RSI Kit™ is a critical component for use in paralytic sedative training and includes all the essentials needed to create multiple scenarios related to intubation when concern for aspiration exists. This PRACTI-RSI Kit is the same as our original, only without needles or syringes.
Each kit contains:
PRACTI-Etomidate 40 mg/20 mL Vial
PRACTI-Atropine 10 mL Prefilled Syringe
PRACTI-Midazolam 10 mg/2 mL Vial
PRACTI-Succinylcholine 20 mg/mL Vial
PRACTI-Ketamine mL Vial
PRACTI-Vecuronium Bromide 10 mg/10 mL Vial
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Specifications
  • Désignation:
    Practi-RSI refill kit