ScrubberStand™ Stackable Stands
Stackable ScrubberStand™, For FlaskScrubber™ Vantage glassware washers
Catalog # 77400-900
Supplier: Labconco
: 4618001
Jump to:
Product Details & Documents
Double your washing capacity by stacking 2 glassware washers, reduce floor space.
- Space efficiency: Optimizes space by enabling vertical stacking of washers
- Increased capacity: Doubles washing capacity without expanding laboratory area
- Safe: ETL certification eliminates any worries over the possibility of tipping
- Ease of installation: Comes already assembled
- Compatibility: Designed to work seamlessly with our existing range of glassware washers
Maximize glassware washing capacity without using extra floor space. Stack 2 Freestanding Washers with a Stackable ScrubberStand™. Made of 1.5” tubular, powder-coated steel for durability and stability. Hardware included. Seismic bracket kit available.
Spécifications
- Désignation:Stackable ScrubberStand™
- For Use With:For FlaskScrubber™ Vantage glassware washers