Transport Tote XL for Samples
Product Details & Documents
High-capacity transport container for laboratory samples, instruments, and valuable products.
- Sturdy and spacious, ideal for transporting a large volume of samples
- Stainless steel, polypropylene, elastomeric compound, ABS construction
- Transparent lid enables effortless identification of contents, reducing the risk of contamination
- Equipped with articulated handles featuring silicone grips for comfortable and secure handling
A reliable solution for the safe and convenient transportation of laboratory samples, instruments, and valuable items. Crafted to meet the stringent demands of clinical and research laboratories, this oversized transport container ensures the protection and preservation of cargo.
A silicone seal ensures a tight enclosure, safeguarding samples from most environmental elements. Dual latches feature a security tag to guarantee a secure closure, providing tamper-evidenct security and ensuring the integrity and authenticity of samples. The bottom compartment is generously sized, allowing for carrying most tube racks, instruments or supplies according to specific requirements. Includes a MultiTube Rack Insert to accommodate various tube sizes. Additionally, two other insert options for blood tubes up to 10ml and general research tubes up to 15 ml are available separately.
Transparent design helps comply with OSHA 29 CFR 1910.1030 standards without having to re-label containers and enabling the user to visibly check sample integrity. This container meets UN3373 secondary packaging transport requirements.
Spécifications
- Capacité:19 L (~5 gal.)
- Color:Red