Microscope Slide Boxes, 25-Place Foam | Avantor
Microscope Slide Boxes, 25-Place Foam
ABS, 25 - Place (26× 76 mm and 1× 3 in slide size), Blue
Catalog # 77395-342
Supplier:   VWR International
Microscope Slide Boxes, 25-Place Foam
Catalog # 77395-342
Supplier:  VWR International
Microscope Slide Boxes, 25-Place Foam
Microscope Slide Boxes, 25-Place Foam
ABS, 25 - Place (26× 76 mm and 1× 3 in slide size), Blue
Price
$19.38
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each Case of 72
Product Details & Documents
Easily access slides with friction-fit cover.
  • Organize and identify; durable and safe
  • Lining and slots are numbered to correspond to the slide inventory sheet
  • Organize by 'Project' and 'Date' with pre-molded labeling area
  • Available in multiple colors for project coding
  • ABS plastic boxes available with either cork or foam lining
  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage
The premium microscope slide box is perfect for the storage or transportation of standard size slides (25×75 mm and 1×3").
Spécifications
  • Capacité:
    25 - Place (26× 76 mm and 1× 3 in slide size)
  • Color:
    Blue
  • Matière:
    ABS