Microscope Slide Boxes, 25-Place Foam
ABS, 25 - Place (26× 76 mm and 1× 3 in slide size), Blue
Catalog # 77395-342
Supplier: VWR International
Jump to:
Product Details & Documents
Easily access slides with friction-fit cover.
- Organize and identify; durable and safe
- Lining and slots are numbered to correspond to the slide inventory sheet
- Organize by 'Project' and 'Date' with pre-molded labeling area
- Available in multiple colors for project coding
- ABS plastic boxes available with either cork or foam lining
- Designed to securely store slides in grooved slots
- Molded feet secure stacking for easy transport and storage
The premium microscope slide box is perfect for the storage or transportation of standard size slides (25×75 mm and 1×3").
Spécifications
- Capacité:25 - Place (26× 76 mm and 1× 3 in slide size)
- Color:Blue
- Matière:ABS