Axygen® LTS Compatible Pipet Tips | Avantor
Axygen® LTS Compatible Pipet Tips
LTS compatible, extended length, 20 µl, +, 49.45 mm, Clear, 10 racks, 96 each
Catalog # 77395-592
Supplier:   Corning
Fournisseur #:  LT-20-TS-S
Axygen® LTS Compatible Pipet Tips
Catalog # 77395-592
Supplier:  Corning
Axygen® LTS Compatible Pipet Tips
Axygen® LTS Compatible Pipet Tips
LTS compatible, extended length, 20 µl, +, 49.45 mm, Clear, 10 racks, 96 each
Price
$138.00
Log in pour la disponibilité
Pack of 960 Case of 4800
Product Details & Documents
The new Axygen® LTS compatible pipet tips are ergonomically designed to reduce discomfort and are optimized to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimized to work with LTS pipettors enables less force to be used during pipetting.
 : ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
Spécifications
  • Volume:
    20 µl
  • Longueur:
    49.45 mm
  • Tip Color:
    Clear
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Tip Style:
    LTS compatible, extended length