Axygen® LTS Compatible Pipet Tips
LTS compatible, extended length, 1000 µl, +, 98.58 mm, Clear, 8 racks, 96 each
  77395-576
 :   Corning
 :  LT-1000-R-S
Axygen® LTS Compatible Pipet Tips
  77395-576
 :  Corning
Axygen® LTS Compatible Pipet Tips
Axygen® LTS Compatible Pipet Tips
LTS compatible, extended length, 1000 µl, +, 98.58 mm, Clear, 8 racks, 96 each
$108.00
Pack of 768 Case of 3840
The new Axygen® LTS compatible pipet tips are ergonomically designed to reduce discomfort and are optimized to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimized to work with LTS pipettors enables less force to be used during pipetting.
 : ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
  • Volume:
    1000 µl
  • Longueur:
    98.58 mm
  • Tip Color:
    Clear
  • Packed:
    8 racks, 96 each
  • Sterile:
    +
  • Tip Style:
    LTS compatible, extended length