Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Microscope Slide Box, 100-Place

Microscope Slide Box, 100-Place

ABS, 100 - Place (26×76 mm and 1×3 in slide size), White
Catalog # 77395-354
Supplier:   VWR International
Microscope Slide Box, 100-Place
Catalog # 77395-354
Supplier:  VWR International
Microscope Slide Box, 100-Place
Microscope Slide Box, 100-Place

ABS, 100 - Place (26×76 mm and 1×3 in slide size), White

Price
$30.26
New Customer Promotional Price
Log in for availability
Each Case of 72
Product Details & Documents

Closes securely with a rust-resistant nickelplated clasp and pin hinges.

  • Organize and identify
  • Lining and slots are numbered to correspond to the slide inventory sheet
  • Organize by 'Project' and 'Date' with pre-molded labeling area
  • Available in multiple colors for project coding
  • Durable and safe
  • ABS plastic boxes available with either cork or foam lining
  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage

The premium microscope slide box is perfect for the storage or transportation of standard size slides.

Documents

Specifications
  • Color:
    White
  • Matière:
    ABS
  • For Use With:
    100 - Place (26×76 mm and 1×3 in slide size)